Order Entry
ContactUsLinkComponent
Anti-PTPRC Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MEM-56]
Anti-PTPRC Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MEM-56]
Catalog #: 89329-712
Supplier:  Genetex
Anti-PTPRC Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MEM-56]
Catalog #: 89329-712
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Tyrosine Phosphatase Receptor Type C
  • Clonality:
    Monoclonal
  • Clone:
    MEM-56
  • Gene ID:
    5788
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Antigen symbol:
    PTPRC
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Size:
    100 test
  • Western blot:
    Yes
  • Antigen synonyms:
    CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
  • Storage buffer:
    Preservative: 15mM Sodium Azide. Constituents: PBS, BSA. pH 7.4
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Immunogen:
    Human thymocytes and T lymphocytes.
  • Purification:
    Protein A purified
  • Cat. no.:
    89329-712
  • Supplier No.:
    GTX18240

Specifications

About this item

FACS: This reagent is designed for Flow Cytometry analysis of human blood cells using 20 ul reagent / 100 ul whole blood. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: MEM-56
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Customers Who Bought This Also Bought