To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Tumor Necrosis Factor Alpha
- Clonality:Polyclonal
- Gene ID:7124
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:TNFA
- Conjugation:Biotin
- ELISA:Yes
- Size:25μg
- Western blot:Yes
- Antigen synonyms:tumor necrosis factor ligand superfamily member 2|tumor necrosis factor alpha|DIF|monocyte-derived|Tnfa|cachectin|Tnfsf1a|TNFalpha|member 2|TNF-alpha|TNF-a|TNFSF2|macrophage-derived|TNF superfamily|TNF|tumor necrosis factor-alpha|APC1 protein
- Storage buffer:Preservative: None. Constituents: 0.1% BSA, PBS
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:0.05 mg/ml
- Immunogen:Highly pure (>98%) recombinant human TNF alpha
- Purification:Immunogen affinity purified
- Cat. no.:89261-842
- Supplier No.:GTX17442
Specifications
About this item
ELISA: Use at a concentration of 0.15 - 0.3 ?g/ml. Used in conjunction with compatible secondary reagents, allows the detection of at least 0.2 ng/well of recombinant human TNF alphA. WB: Use at a concentration of 0.1 - 0.2 ?g/ml. Predicted molecular weight: 26 kDa. Used in conjunction with compatible secondary reagents the detection limit for recombinant human TNF alpha is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: TNFA
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Human