Order Entry
ContactUsLinkComponent
Anti-ABCC11 Mouse Monoclonal Antibody [clone: 2Q1841]
Anti-ABCC11 Mouse Monoclonal Antibody [clone: 2Q1841]
Catalog #: 89287-920
Supplier:  Genetex
Anti-ABCC11 Mouse Monoclonal Antibody [clone: 2Q1841]
Catalog #: 89287-920
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ATP-binding cassette, Subfamily C (CFTR/MRP), Member 11
  • Clonality:
    Monoclonal
  • Clone:
    2Q1841
  • Gene ID:
    6279
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Antigen symbol:
    ABCC11
  • Conjugation:
    Unconjugated
  • ImmunoFluorescence:
    Yes
  • Size:
    50 μg
  • Western blot:
    Yes
  • Antigen synonyms:
    WW|member 11|MRP8|sub-family C (CFTR/MRP)|ATP binding cassette subfamily C member 11|ATP-binding cassette|EWWD
  • Storage buffer:
    Preservative: 0.1% Sodium Azide; Constituents: PBS. pH 7.2
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Neutrophils
  • Purification:
    Purified by affinity chromatography.
  • Cat. no.:
    89287-920
  • Supplier No.:
    GTX14576

Specifications

About this item

ICC: Use at an assay dependent dilution (ethanol fixed cells)WB: Use at an assay dependent dilution. Use under non reducing conditions. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ABCC11
Clonality: Monoclonal
Clone: 2Q1841
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Customers Who Bought This Also Bought