Order Entry
ContactUsLinkComponent
Anti-DUT Rabbit Polyclonal Antibody
Anti-DUT Rabbit Polyclonal Antibody
Catalog #: 89278-824
Supplier:  Genetex
Anti-DUT Rabbit Polyclonal Antibody
Catalog #: 89278-824
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    dUTPase
  • Clonality:
    Polyclonal
  • Gene ID:
    1854
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Antigen symbol:
    DUT
  • Conjugation:
    Unconjugated
  • Size:
    100 μg
  • Western blot:
    Yes
  • Antigen synonyms:
    Deoxyuridine 5'-triphosphate nucleotidohydrolase|DUT|mitochondrial|dUTP pyrophosphatase
  • Storage buffer:
    Preservative: 0.02% Sodium Azide; Constituents: PBS
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    0.5 mg/ml
  • Immunogen:
    Synthetic peptide, corresponding to amino acids 25-44 (NARGTAEGRSRGTLRARPAP) and 113-130 (QLRFARLSEHATAPTRGS) of Human DUT-N.
  • Purification:
    Protein G purified
  • Cat. no.:
    89278-824
  • Supplier No.:
    GTX13690

Specifications

About this item

WB: Use at a dilution of 1/1000 - 1/2000. Detects a band of approximately 21 kDa (predicted molecular weight: 27 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: DUT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Customers Who Bought This Also Bought