To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor 132
- Clonality:Polyclonal
- Gene ID:29933
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:GPR132
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:25μg
- Antigen synonyms:Probable G-protein coupled receptor 132.|GPR132|G2 accumulation protein|G2A|G Protein Coupled Receptor G2A|GP132_HUMAN
- Storage buffer:Preservative: 0.01% Sodium Azide; Constituents: PBS. pH 7.7
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide (unfortunately, the amino acid sequence is considered to be commercially sensitive)
- Purification:Immunogen affinity purified
- Cat. no.:89277-602
- Supplier No.:GTX12530
Specifications
About this item
IHC-P: Use at a dilution of 1/100 - 1/200. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: GPR132
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human