To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Actin F
- Clonality:Monoclonal
- Clone:AE-7
- Gene ID:60
- Host:Mouse
- Isotype:IgM
- Reactivity:Human
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:2ml
- Antigen synonyms:PS1TP5BP1|BRWS1
- Storage buffer:Preservative: 0.09% sodium azide; Constituents: Tissue Culture Supernatant.
- Storage temperature:For long term storage freeze working aliquots at –20 °C in a manual defrost freezer. Avoid Repeated Freeze Thaw Cycles.
- Immunogen:This antibody recognizes an antigen associated with actin bundles found in human cells.
- Purification:Tissue culture supernatant
- Cat. no.:89280-384
- Supplier No.:GTX12360
Specifications
About this item
IHC-P: Use at an assay dependent dilution. IHC-Fr: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: [delete]
Clonality: Monoclonal
Clone: AE-7
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human