To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Polo like kinase 1
- Clonality:Monoclonal
- Clone:AZ27
- Gene ID:380481
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Xenopus,Human,Mouse
- Antigen symbol:PLK1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:polo-like kinase|Serine/threonine-protein kinase 13|Polo-Like Kinase pS482/pS486/pS490,STPK13|PLK-1|polo-like kinase 1
- Storage buffer:Preservative: NoneConstituents: PBS
- Concentration:0.4 mg/ml
- Immunogen:Recombinant full length protein (Xenopus laevis).
- Purification:Protein A purified
- Cat. no.:89328-234
- Supplier No.:GTX12210
Specifications
About this item
ELISA: Use at an assay dependent dilution. IHC: Use at an assay dependent dilution. WB: Use at a concentration of 1 ?g/ml. Detects a band of approximately ~55 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: plk1
Clonality: Monoclonal
Clone: AZ27
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Xenopus laevis