To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Kaiso
- Clonality:Monoclonal
- Clone:2T15
- Gel shift:Yes
- Gene ID:56805
- Host:Mouse
- Isotype:IgG1
- Reactivity:Hamster,Human,Dog,Rat,Mouse
- Antigen symbol:Zbtb33
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:50 μg
- Western blot:Yes
- Storage buffer:Preservative: 0.05% Sodium Azide; Constituents: 30% Glycerol, 0.15M Sodium Chloride, 0.1M Tris-glycine. pH 7.4
- Storage temperature:stored at 4 °C for short-term only. For long-term storage, store at –20 °C. Aliquots are stable for at least 12 months at –20 °C.
- Concentration:1 mg/ml
- Immunogen:Recombinant fusion protein (N-terminal), corresponding to amino acids 103 - 584 of mouse Kaiso.
- Purification:Protein G purified
- Cat. no.:89317-312
- Supplier No.:GTX11810
Specifications
About this item
For GSA: Use at an assay dependent dilution. Supershifts a specific methyl-CpG complex. For IHC: Use at a concentration of 1 ?g/ml for positive nuclear staining in NIH3T3 cells fixed with 100% methanol. For IP: Use at an assay dependent dilution. For WB: Use at a concentration of 0.5 - 2 ?g/ml on RIPA lysates from 3T3/A31 cells (ECL). Detects a band of approximately 97 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Zbtb33
Clonality: Monoclonal
Clone: 2T15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Dog, Rat, Hamster