To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Matrix Metalloproteinase 21
- Clonality:Polyclonal
- Gene ID:118856
- Host:Rabbit
- Isotype:IgG
- Reactivity:Xenopus,Human,Rat,Mouse
- Antigen symbol:MMP21
- Conjugation:Unconjugated
- Size:50 μg
- Western blot:Yes
- Antigen synonyms:MMP-21
- Storage buffer:Preservative: 0.05% Sodium Azide; Constituents: 50% Glycerol, PBS
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide (Human) Hinge.
- Purification:Immunogen affinity purified
- Cat. no.:89287-862
- Supplier No.:GTX11637
Specifications
About this item
WB: Use at an assay dependent dilution. A minimum working antibody dilution of 1/1000 is determined by blotting a tissue cell lysate with an alkaline phosphatase conjugated secondary antibody and BCIP/NBT as the substrate. A starting dilution of 1/5000 of GTX11637 is recommended for chemiluminescent substrates. By blotting against the reduced protein, the antibody identifies bands at 70 kDa and 66 kDa. This is larger than the predicted 65 kDa and possibly represents alternative splicing, post translational modifications, aggregation, or other phenomenon. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: MMP21
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Xenopus laevis,