To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Ribonucleic Acid Export 1
- Clonality:Polyclonal
- Gene ID:9869
- Host:Rabbit
- Isotype:IgG
- Reactivity:Pig,Human,Bovine,Dog,Rat,Mouse
- Antigen symbol:RAE1
- Conjugation:Unconjugated
- ImmunoPrecipitation:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:mRNA-associated protein MRNP 41|MIG14|MGC126076|OTTHUMP00000166110|RP4-800J21.2|mRNA-binding protein|dJ481F12.3|mRNA export protein|migration-inducing gene 14|MGC126077|MRNP41|HGNC:9828|RAE1|Mnrp41|S.pombe) homolog|homolog of yeast Rae1 (Bharathi) mRNA-associated protein of 41 kDa (Kraemer)|MGC117333|FLJ30608|41-kD|dJ800J21.1|RAE1 RNA export 1 homolog (S. pombe) |RAE1 (RNA export 1
- Storage buffer:1XPBS, 1% BSA, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human SETDB1. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89322-106
- Supplier No.:GTX115305
Specifications
About this item
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IP: 1:100-1:500, ChIP. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: RAE1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Pig, Rat