To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Galanin Receptor 3
- Clonality:Polyclonal
- Gene ID:1052
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:GALR3
- Conjugation:Unconjugated
- Size:100 μl
- Antigen synonyms:NF-IL6-beta|C/EBP-delta|CELF|CRP3
- Storage buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0.93 mg/ml
- Immunogen:Full length human C/EBP delta Recombinant protein.
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89312-592
- Supplier No.:GTX115047
Specifications
About this item
WB,ICC/IF:Use at an dependent assay.Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: CEBPD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse