To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Glioma Tumor Suppressor Candidate Region 2
- Clonality:Monoclonal
- Clone:HEK/1/85a
- Gene ID:1234
- Host:Rat
- Isotype:IgG2a
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Antigen symbol:GLTSCR2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Size:100 μg
- Antigen synonyms:protein interacting with carboxyl terminus 1.|GSCR2_HUMAN|GLTSCR2|p60|PICT1|Glioma tumor suppressor candidate region gene 2 protein
- Storage buffer:Phosphate buffered saline containing 0.09% Sodium Azide
- Storage temperature:Store at +4 °C or at –20 °CThis product should be stored undiluted. Storage in frost-free freezers is not recommended. Should this product contain a precipitate we recommend microcentrifugation before use.
- Immunogen:CHO cells transfected with human CCR5.
- Purification:Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
- Cat. no.:89285-156
- Supplier No.:GTX11464
Specifications
About this item
Recommended Starting Dilutions:Flow Cytometry: 1/50 - 1/100Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: GLTSCR2
Clonality: Monoclonal
Clone: HEK/1/85a
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Rat