To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Tubulin, Beta 3 Class Iii
- Clonality:Monoclonal
- Clone:SDL.3D10
- Gene ID:10381
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Antigen symbol:E2F7
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:CDCBM|beta-4|CFEOM3A|FEOM3|CDCBM1|TUBB4|CFEOM3
- Storage buffer:Preservative: 15mM Sodium Azide; Constituents: Raw Ascites
- Storage temperature:Aliquot and store at –20 °C or –80 °C. Avoid repeated freeze / thaw cycles
- Immunogen:Chemically synthesized peptide corresponding to the carboxyl-terminal sequence of human beta-tubulin isotype III conjugated to BSA.
- Purification:Ascites
- Cat. no.:89316-558
- Supplier No.:GTX11314
Specifications
About this item
Mouse monoclonal antibody [SDL.3D10] to beta III Tubulin
Indirect ELISA: Use at an assay dependent dilution. WB: Use at a dilution of 1/400. Predicted molecular weight: 55 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: E2F7
Clonality: Monoclonal
Clone: SDL.3D10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human