To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Discs, Large (Drosophila) Homolog-Associated Protein 4
- Clonality:Polyclonal
- Gene ID:3567
- Host:Goat
- Isotype:IgG
- Neutralisation:Yes
- Reactivity:Human
- Antigen symbol:DLGAP4
- Conjugation:Unconjugated
- ELISA:Yes
- Size:250 μg
- Western blot:Yes
- Antigen synonyms:DAP-4|DLP4|SAPAP-4|SAPAP4|DAP4|discs large homolog associated protein 4
- Storage buffer:Preservative: NoneConstituents: PBS
- Concentration:1mg/ml
- Immunogen:Recombinant, human interleukin 5, expressed in Sf 21 insect cells.
- Purification:Protein G purified
- Cat. no.:89317-236
- Supplier No.:GTX10757
Specifications
About this item
Goat polyclonal to IL5
ELISA: Use at a concentration of 1 ?g/ml to detect 1 ng/well of recombinant, human IL5. Neut: Use at a concentration of 5.8 ?g/ml. WB: Use at a concentration of 2 mg/ml to detects 5 ng/lane of human IL5 under reducing and non-reducing conditions. Predicted molecular weight: 34 kDa. Anti-Human IL5 was tested for its ability to neutralize the biological activity of recombinant, human IL5 on human TF-1 cells, adapted to grow with IL5 stimulation. The ND50 of the antibody is defined as the concentration of antibody resulting in a one-half maximal inhibition of bioactivity of recombinant, human IL5, when IL5 is present at a concentration just high enough to elicit a maximum response. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: DLGAP4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human