To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Methylthioribose-1-Phosphate Isomerase 1
- Clonality:Polyclonal
- Gene ID:2903
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:MRI1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:Translation Initiation Factor EIF-2B Subunit Alpha/Beta/Delta-Like Protein|Ypr118w|Mediator Of RhoA-Dependent Invasion|MRDI|MTNA
- Storage buffer:1XPBS, 1% BSA, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the N-terminus region of human NMDAR2A. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89319-002
- Supplier No.:GTX103558
Specifications
About this item
Rabbit Polyclonal antibody to NMDAR2A (glutamate receptor, ionotropic, N-methyl D-aspartate 2A)
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: MRI1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human