To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Proteasome 26S Subunit, ATPase 3
- Clonality:Monoclonal
- Clone:11F5
- Gene ID:7037
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Monkey,Human,Dog,Rat
- Antigen symbol:PSMC3
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Antigen synonyms:TBP1|macropain) 26S Subunit|proteasome 26S subunit|Proteasome (prosome|ATPase 3
- Storage buffer:Preservative: 0.1% Sodium Azide; Constituents: PBS. pH 7.4
- Immunogen:Recombinant full length protein (Human).
- Purification:Protein A purified
- Cat. no.:89336-518
- Supplier No.:GTX10249
Specifications
About this item
Mouse monoclonal antibody [11F5] to CD71
ELISA: Use at an assay dependent dilution. Recommended pair antibody: GTX10249 (detection) with GTX10252 (coating). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: PSMC3
Clonality: Monoclonal
Clone: 11F5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Dog, Monkey, Rat