To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Tubulin Tyrosine Ligase-like Family Member 12
- Clonality:Polyclonal
- Gene ID:6494
- Host:Goat
- Isotype:IgG
- Reactivity:Monkey,Human,Dog,Rat
- Antigen symbol:TTLL12
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:EDG2|fSAP17|EDG-2|G10|Cwc14|YCR063W
- Storage buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:C-ASKQLGSPTADLA
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:89287-842
- Supplier No.:GTX10162
Specifications
About this item
Goat polyclonal antibody to SPA1 / SIPA1
Approx. 150kDa band observed in Human lysates of EBV-immortilized lymphoblastoid (calculated MW of 112kDa according to NP_694985.28 and NP_006738.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Kurachi et al, J Biol Chem. 1997 Oct 31;272(44):28081-8; PMID: 9346962). Recommended concentration: 1-3µg/ml. An additional band of 55-60kDa was consistently observed, however this band was not blocked by the immunizing peptide and it is therefore a non-specific signal. We call for caution when this product is used for other assays than Western blot. Both variants represent identical protein (NP_694985.28 and NP_006738.3).
Type: Primary
Antigen: TTLL12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Dog, Monkey, Rat