To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD4 molecule
- Clonality:Monoclonal
- Clone:11-60
- Gene ID:3565
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:CD4
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Antigen synonyms:L3T4|p55|CD4mut|Ly-4|W3/25
- Storage buffer:Preservative: 0.1% Sodium Azide; Constituents: PBS. pH 7.4
- Immunogen:Recombinant full length protein (Human).
- Purification:Protein G purified
- Cat. no.:89336-480
- Supplier No.:GTX10107
Specifications
About this item
Mouse monoclonal [11 - 60] to IL4
ELISA: Use at an assay dependent dilution. Can be used in antibody pairs with GTX10105 and 10106 (both capture). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 20759
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: