KOGO™ TPR Cleanroom Tape
Supplier: Texwipe
New Product
Certificates
About this item
These tapes are constructed of low density polyethylene backing with rubber adhesive
- Easy identification, protection and storage
- Reduces need for cutting or splicing tape with available width options
- Holds to irregular shapes and creates proper seal/protection
- Aggressive tack with quick bond
These tapes are available in 5 colors and 4 widths. Also they are ideal for color coding or product identification in cleanrooms. The highly conformable tape makes it well suited for sealing compound surfaces and packaging materials within critical environments.
This construction is low in halogens, leachable chlorides and heavy metals and provides users with the ideal cleanroom tape solution where quick tack but moderate adhesion is needed.
Ordering information: Tapes are available in press-and-seal cleanroom bags and also gamma irradiated tapes are available.
Customers Who Bought This Also Bought