Sharps Bank Waste Disposal Unit
Catalog #: 10796-894
Supplier: Whitney Products
Specifications
- Capacity:45.4 L (12 gal.)
- Cat. no.:10796-894
- Supplier No.:SB1201
Specifications
About this item
These containers are designed for the disposal of contaminated items in the lab environment.
They feature 3 walled construction: Inner and outer carton provide puncture resistance; a 3 mil liner provides leak resistance. They have an extra wide opening for safe, convenient disposal and the two-flap lid configuration protects contents from spilling. The containers have a square bottom to reduce the chance of tipping and the two handle construction makes transporting these containers to central disposal convenient. Containers ship and store flat for space-saving storage.
Customers Who Bought This Also Bought