KOGO™ Cleanroom Tapes, TVR Series, Texwipe®
Supplier: Avantor
Certificates
About this item
Tape is made of polyvinyl chloride backing with synthetic rubber adhesive and is latex-free.
- Tapes available in five colors and four widths
- With labeling and write on area
- Bond with irregular surfaces like ductwork sealing and visqueen sheets
- HEPA filter protection
These tapes are ideal for clean build or cleanroom construction applications where quick tack and abrasion resistance is important.
Ordering information: Tapes are available in press-and-seal cleanroom bags.
Customers Who Bought This Also Bought