Premium Two-Prong Clamp, Cork Lined
Supplier: Wards
Certificates
About this item
Cork Lined Extension Lab Clamp
- Maximum clamp opening is 11 cm
- 14 cm connecting rod.
Clamp constructed of powder coated steel, two cork lined prongs attached to an aluminum rod. Completely metal construction with thread fitted actuating joints and 10 mm diameter connecting rod.