Premium Four-Prong Clamp, Vinyl Coated
Supplier: Wards
Certificates
About this item
Vinyl Lined Extension Lab Clamp
- Maximum clamp opening 10.5 cm
- 14 cm connecting rod
Clamp constructed of powder coated steel, four vinyl coated prongs attached to an aluminum rod. Completely metal construction with thread fitted actuating joints and 10 mm diameter connecting rod.