DuPont™ Tyvek® IsoClean® Boot Covers with Gripper™ Sole
Supplier: DuPont
Certificates
About this item
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
- Gripper™ soles provide enhanced skid-resistance and durability
- Elastic at ankles allows for a secure fit
- Elastic opening
- Coverage up to calf
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.
Certifications: Sterile garments are gamma sterilized to SAL of 10-6.
Packaging: Bulk packaged option packed in double transparent poly liners.
Specifications
- Catalog No:
- 10016-088
- CA10016-098
- Garment application:
- Cleanroom
- Cleanroom