Specifications
- Description:Ward's® Chemistry Acetylene-Chlorine Reaction Demonstration
- No. of Lab Groups:8
- Cat. no.:470010-830
- Includes:Instructions
- Supplier No.:WARD470010-830
Specifications
About this item
Study exothermic and oxidation-reduction reactions with ease.
- Aligned to National Science Education standards
- This demonstration can be performed eight times
This reaction demonstrates the reaction of acetylene and chlorine to form hydrochloric acid and carbon. This dramatic demonstration of a spontaneous reaction also generates light as a product.