Specifications
- Description:Ward's® Formation Of Esters Lab Activity
- No. of Lab Groups:8
- Cat. no.:470018-102
- Includes:Instructions, Student Guide
- Supplier No.:WARD470018-102
Specifications
About this item
Synthesize a Variety of Esters by Reactions of Organic Acids and Alcohols
- Introduces students to simple organic synthesis reactions
- Allows students practice stoichiometry skills
- Takes one 50 minute class period to complete
The reaction that is performed in this lab activity is commonly referred to as a Fischer esterification reaction. Esters are compounds that frequently have distinctive odors and are naturally occurring flavor and fragrance chemicals in many fruits and plants. In this lab, the smells of the esters produced will be investigated.