Finntip®, Special Pipette Tips, Thermo Scientific
Catalog #: 14233-396
Supplier: Thermo Fisher Scientific
Specifications
- Volume:100 - 1000 µL
- Tip Color:Blue
- Packed:5 Racks, 96 each
- Sterile:+
- Supplier No.:9401423
Specifications
About this item
Finntip® Extended are manufactured from high quality polypropylene.
- Autoclavable
These long tapered tip especially adapted for deep tubes and narrow-necked bottles.
Customers Who Bought This Also Bought