Ward's® Half Life Prop Radioactive Element
Catalog #: 470023-826
Supplier: Avantor
Specifications
- Description:Wards Half Life Prop Radioactive Element
- No. of Lab Groups:15
- Cat. no.:470023-826
- Supplier No.:WARD470023-826
Specifications
About this item
This is a great experiment to study half life and radioacticve elements in the classroom!
- Includes enough materials for 15 setups, a teacher's guide, and student copymasters
Radioisotopes have been found to be useful in the fields of geology, medical diagnosis and treatment, metallurgy, agriculture, and scientific research. This lab allows students to see a demonstration of nuclear decay using a model of uranium-238. By graphing their data, they better understand the reason for radioactive decay and the concepts of subatomic particles, half life, and more.
Customers Who Bought This Also Bought