To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Glutathione Peroxidase 1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Antigen symbol:GPX1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Epitope:amino acids 175-194 of Human glutathione peroxidase 1 ( GPx-1)
- Cross adsorption:No
- Format:Lyophilized
- Antigen synonyms:GSHPX1|GSHPx-1|Cellular glutathione peroxidase
- Immunogen:A synthetic peptide (RRYSRRFQTIDIEPDIEALL) corresponding to the amino acids 175-194 of human glutathione peroxidase 1 ( GPx-1) conjugated to diphtheria toxin has been used as the immunogen.
- Cat. no.:10782-934
- Supplier No.:R-1588-100
Specifications
About this item
Glutathione peroxidase 1 has a role in detoxification of hydrogen peroxide and is one of the most important antioxidant enzymes in humans. It exists as a homotetramer which localises to the cytoplasm. It belongs to the glutathione peroxidase family. Glutathione peroxidase 1 is one of few proteins in higher vertebrates to contain selenocysteine, which occurs at the active site of glutathione peroxidase and is coded by UGA, that normally functions as a translation termination codon. This protein has a polyalanine sequence polymorphism in the N-terminal region, which includes three alleles with five, six or seven alanine repeats. The allele with five alanine repeats is significantly associated with breast cancer risk. At least two alternatively spliced isoforms have been identified.
Human and rat cellular selenium dependent glutathione peroxidase.
Application Information:
IHC, WB and ELISA. This antibody works superbly in IHC on frozen or wax embedded tissues. Antigen retrieval has been used in testing but may not be necessary. Typical working dilutions for routine IHC are 1: 200 to 1: 1,000 depending on tissue and detection method. For WB a dilution range of 1: 1,000 to 1: 4,000 is recommended. For ELISA, a dilution of 1: 1,000 to 1: 4,000 is recommended. This antiserum has extremely high titre, it stains human and rat glial cells intensely and to some extent also stains neurons. Other tissues have not yet been tested. On WB under reducing conditions recognises a 22kD protein in human brain tissue and Sigma glutathione peroxidase (G-4013) isolated from human erythrocytes. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: glutathione peroxidase 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: amino acids 175-194 of Human glutathione peroxidase 1 ( GPx-1)
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat,