To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Neuron Specific Enolase
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:NSE
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:50 µL
- Western blot:Yes
- Epitope:Multiple points of reactivity likely because of the polyclonal nature of the antibody
- Cross adsorption:No
- Format:Lyophilized
- Antigen synonyms:Eno|Neuron Specific Enolase|ENOG|Neuronal Enolase|Neural Enolase|glycerate hydrolyase
- Immunogen:Recombinant human Neuron Specific Enolase (NSE) expressed in and purified from E.coli
- Cat. no.:10782-924
- Supplier No.:R-1396-50
Specifications
About this item
Enolase is a metalloenzyme that catayzes the reaction between 2-phospho-D-glycerate and phosphoenolpyruvate during glycolysis. Mammalian enolase is composed of 3 subunits; alpha, beta and gamma (Neuron-specific enolase). These subunits can form homodimers or heterodimers. The alpha/gamma heterodimer and the gamma/gamma homodimer are found primarily in neurons.
Specifically recognizes ~47 kDa NSE protein in WB.
Application Information:
Western Blotting (WB) and Immunocytochemistry (ICC). A dilution of 1:1,000 - 1:2,000 is recommended for WB. A dilution of 1:500 is recommended for ICC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Neuron specific enolase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Mouse