To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Adaptor Protein, Phosphotyrosine Interaction, PH Domain And Leucine Zipper Containing 2
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Mouse
- Antigen symbol:APPL2
- Conjugation:Unconjugated
- ImmunoPrecipitation:Yes
- Size:100 µL
- Western blot:Yes
- Epitope:Mouse APPL2 protein (amino acids: 619-662)
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:DIP13B
- Immunogen:Mouse APPL2 protein (amino acids: 619-662) conjugated to GST.
- Cat. no.:10782-718
- Supplier No.:R-1332-100
Specifications
About this item
APPL2 is an isoform of APPL1 with 54% identity in their protein sequences. APPL2 has a role in cell proliferation and embryonic development. Recently, it was shown that APPL2 regulates FSH signaling and acts as a negative regulator in adiponectin signaling.
- Antibody is known to recognize APPL2 by Western Blotting (WB) and Immunoprecipitation (IP). A dilution of 1:1,000 is recommended for WB. A concentration of 3 µl/reaction is recommended for IP. Biosensis recommends optimal dilutions/concentrations should be determined by the end user. This antiserum is known to recognise both mouse and human APPL2.
- Application Information:
- Western Blotting (WB) and Immunoprecipitation (IP). A dilution between 1:2500 and 1:5000 is recommended for WB. A concentration of 3 µl/reaction is recommended for IP. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: APPL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Mouse APPL2 protein (amino acids: 619-662)
Host: Rabbit
Isotype:
Reactivity: Mouse, Human