To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Brain-Derived Neurotrophic Factor
- Clonality:Polyclonal
- Host:Sheep
- Isotype:IgG
- Neutralisation:Yes
- Reactivity:Human,Rat,Mouse
- Antigen symbol:BDNF
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 µL
- Western blot:Yes
- Cross adsorption:No
- Format:Lyophilized
- Antigen synonyms:Brain-derived neurotrophic factor|proBDNF|Abrineurin
- Immunogen:Recombinant human BDNF
- Cat. no.:10782-618
- Supplier No.:S-016-100
Specifications
About this item
BDNF belongs to the neurotrophin family and promotes the survival of neuronal populations that are all located either in the central nervous system or directly connected to it. It is a major regulator of synaptic transmission and plasticity at adult synapses in many regions of the CNS. The versatility of BDNF is emphasized by its contribution to a range of adaptive neuronal responses including long-term potentiation (LTP), long-term depression (LTD), certain forms of short-term synaptic plasticity, as well as homeostatic regulation of intrinsic neuronal excitability. The alterations in BDNF expression induced by various kinds of brain insult including stress, ischemia, seizure activity and hypoglycemia, may contribute to some pathologies such as depression, epilepsy, Alzheimer's, and Parkinson's disease. Microglia release BDNF that may contribute to neuroinflammation and neuropathic pain. SUBUNIT: Monomers and homodimers. Binds to NTRK2/TRKB. SUBCELLULAR LOCATION: Secreted protein. POst translation modification: Converted into mature BDNF by plasmin (PLG). SIMILARITY: Belongs to the NGF-beta family.
A cross reactivity of less than 1% against mouse NGF, recombinant human NT3 or NT4/5 has been shown by one site ELISA.
Application Information:
IHC, ELISA, Western Blot, inhibition of biological activity in vitro/in vivo. Recommended to be used at a dilution of 1:200-2000 for immunohistochemistry, ELISA and Western blot. 1:10 to 1:50 for inhibition of biological activity in vitro. Use neat for in vivo studies at 5-10 µl/g body weight. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: rh BDNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: [delete]
Host: Sheep
Isotype: IgG
Reactivity: Rat, Mouse, Human