To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Glutathione Peroxidase 3
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Antigen symbol:GSHPx
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µL
- Epitope:amino acids 135-154 of Human glutathione peroxidase 3 (GPx-P)
- Cross adsorption:No
- Format:Lyophilized
- Antigen synonyms:GSHPx-3|GPx-P|GSHPx-P
- Immunogen:A synthetic peptide (FEKGDVNGEKEQKFYTFLKN) corresponding to the amino acids 135-154 of human glutathione peroxidase 3 (GPx-P) conjugated to diphtheria toxin has been used as the immunogen. The peptide is homologous with the corresponding sequence derived from glutathione peroxidase 3 (GPx-P) protein in mouse, rat, orangutan and bovine.
- Cat. no.:10782-584
- Supplier No.:R-171-100
Specifications
About this item
GPx-P belongs to the glutathione peroxidase family which are responsible for the detoxification of hydrogen peroxide. It protects cells and enzymes from oxidative damage, by catalyzing the reduction of hydrogen peroxide, lipid peroxides and organic hydroperoxide, by glutathione. GPx-P is secreted in plasma and exists as a homotetramer.
This antibody has been shown to be specific for glutathione peroxidase 3 protein (GPx-P) through staining and blocking studies.
Application Information:
IHC, only. A dilution o f 1: 1000 to 1: 5000 is recommended for immunohistochemistry depending on detection method but end user will need to determine dilution to suit application. Antigen retrieval is recommended. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: glutathione peroxidase 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: amino acids 135-154 of Human glutathione peroxidase 3 (GPx-P)
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat