Order Entry
ContactUsLinkComponent
Anti-Nicastrin, C-terminal domain Rabbit Polyclonal Antibody
Anti-Nicastrin, C-terminal domain Rabbit Polyclonal Antibody
Catalog #: 10782-562
Supplier:  Biosensis
Antibody
Anti-Nicastrin, C-terminal domain Rabbit Polyclonal Antibody
Catalog #: 10782-562
Supplier:  Biosensis
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nicastrin
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Antigen symbol:
    NCSTN
  • Conjugation:
    Unconjugated
  • ImmunoPrecipitation:
    Yes
  • Size:
    500 µg
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Antigen synonyms:
    ATAG1874|Nicastrin|KIAA0253|NCSTN|UNQ1874/PRO4317
  • Immunogen:
    A synthetic peptide (C-NAKADVLFIAPREPGAVSY) corresponding to human Nicastrin [691-709] in the C-terminal region conjugated via additional N-terminal Cys to Diphtheria toxoid.
  • Cat. no.:
    10782-562
  • Supplier No.:
    R-1684-500

Specifications

About this item

Nicastrin, a type 1 membrane glycoprotein, is an essential component of the gamma secretase complex which is critical for the cleavage of the amyloid precursor protein and other membrane proteins. Nicastrin is widely expressed in different tissue types. This antibody detects all processed forms of Nicastrin.

Confirmed by WB using peptide absorption.
Application Information:
WB and IP. Suggested concentration of 3-10 ug/mL is recommended for WB. Human or mouse brain samples commonly prepared with reducing (50mM DTT), urea (2.3 M), SDS (1.5%) in 62.5 mM Tris-HCL pH 6.8 sample buffer (without boiling) heating to 50 C for 15 min. Unprocessed full length human Nicastrin is 709 amino acids, however this protein contains an N-terminal signal peptide which is considered to undergo cleavage during processing and transit to the cell plasma membrane, in addition the protein undergoes glycosylation to produce a glycoprotein of about 145 kDa apparent MW by SDS PAGE. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Nicastrin, C-terminal domain
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human Nicastrin [691-709]
Host: Rabbit
Isotype: IgG
Reactivity:

Customers Who Bought This Also Bought