To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Superoxide Dismutase 1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:SOD1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µL
- Western blot:Yes
- Epitope:amino acids 121-139 of Human superoxide dismutase (SOD1)
- Cross adsorption:No
- Format:Lyophilized
- Antigen synonyms:SOD1|ALS|ALS1|Cu/Zn superoxide dismutase|Homodimer|IPOA|Indophenoloxidase A
- Immunogen:A peptide (HEKADDLGKGGNEESTKTG) corresponding to the amino acids 121-139 of human superoxide dismutase (SOD1) conjugated to diphtheria toxin has been used as the immunogen.
- Cat. no.:10782-554
- Supplier No.:R-168-100
Specifications
About this item
SOD1 binds copper and zinc ions ans is one of two isozymes responsible for destroying free superoxide radicals which are normally produced within the cells and which are toxic to biological systems. SOD1 is a soluble cytoplasmic protein, acting as a homodimer to convert superoxide radicals to molecular oxygen and hydrogen peroxide. Defects in SOD1 are the cause of amyotrophic lateral sclerosis type 1 (ALS1) which is a neurodegenerative disorder affecting upper and lower motor neurons and resulting in fatal paralysis.
This antibody has been shown to be specific for superoxide dismutase (SOD1) protein.
Application Information:
IHC, WB. This antibody works superbly in immunohistochemistry on frozen or paraffin embedded tissues. Antigen retrieval has been used in testing but may not be necessary. Typical working dilutions for routine immunohistochemistry are 1: 100 to 1: 1000 depending on tissue and detection method. For western blotting a dilution range of 1: 1000 to 1: 4000 is recommended. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: superoxide dismutase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: amino acids 121-139 of Human superoxide dismutase (SOD1)
Host: Rabbit
Isotype: IgG
Reactivity: Human