To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:AP1 Complex Subunit beta-1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Antigen symbol:AP1B1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µL
- Western blot:Yes
- Epitope:amino acids 11-29 of Human AP-1
- Cross adsorption:No
- Format:Lyophilized
- Immunogen:A synthetic peptide (C-KKGEIFELKAELNSDKKEK) corresponding to the amino acids 11-29 of human AP-1 complex subunit beta-1 conjugated to diphtheria toxin has been used as the immunogen. The peptide is homologous with the corresponding sequence derived from AP-1 complex subunit beta-1 protein in mouse, rat and bovine.
- Cat. no.:10782-546
- Supplier No.:R-167-100
Specifications
About this item
FUNCTION: Subunit of clathrin-associated adaptor protein complex 1 that plays a role in protein sorting in the late-Golgi/trans-Golgi network (TGN) and/or endosomes. The AP complexes mediate both the recruitment of clathrin to membranes and the recognition of sorting signals within the cytosolic tails of transmembrane cargo molecules. SUBUNIT: Adaptor protein complex 1 (AP-1) is an heterotetramer composed of two large adaptins (gamma1/AP1G1 or gamma2/AP1G2 and beta1A/AP1B1 or beta1B/AP1B1), a medium adaptin (mu1A/AP1M1 or mu1B/AP1M2) and a small adaptin (sigma1A/AP1S1 or sigma1B/AP1S2 or sigma1C/AP1S3). SUBCELLULAR LOCATION: Golgi apparatus. Cytoplasmic vesicle; cytoplasmic vesicle membrane; peripheral membrane protein; cytoplasmic side. Note=Component of the coat surrounding the cytoplasmic face of coated vesicles located at the Golgi complex. ALTERNATIVE PRODUCTS: 2 named isoforms produced by alternative splicing. TISSUE SPECIFICITY: Widely expressed. DISEASE: Deletion of the AP1B1 gene may play a role in the tumorigenesis of meningiomas. SIMILARITY: Belongs to the adaptor complexes large subunit family.
This antibody has been shown to be specific for AP-1 complex subunit beta-1.
Application Information:
IHC, WB. This antibody works superbly in immunohistochemistry on frozen or paraffin embedded tissues. Antigen retrieval has been used in testing but may not be necessary. Typical working dilutions for routine immunohistochemistry are 1: 1000 to 1: 5000 depending on tissue and detection method. For western blotting a dilution range of 1: 100 to 1: 10000 is recommended. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: AP1B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: amino acids 11-29 of Human AP-1
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat