To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:beta Lipotropin
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:beta Lipotropin
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:50 µg
- Epitope:Multiple epitopes because of the nature of the immunogen
- Cross adsorption:No
- Format:Lyophilized
- Antigen synonyms:Beta-LPH|Lipotropin beta|POMC|Pro-opiomelanocortin
- Immunogen:Synthetic peptide (179-267 aa) of human beta-Lipotropin conjugated to thyroglobulin was used as the immunogen.
- Cat. no.:10782-488
- Supplier No.:R-1493-50
Specifications
About this item
Human beta-Lipotropin is a 93 amino acid polypeptide that is cleaved from carboxy-terminal fragment of the precursor pro-opiomelanocortin (POMC). It stimulates melanocytes to produce melanin, and can also be cleaved into smaller peptides including opioid peptides: gamma-lipotropin, alpha-MSH, beta-MSH, gamma-MSH, alpha-endorphin, beta-endorphin, gamma-endorphin and met-enkephalin
Antibody recognizes beta Lipotropin by native applications. A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Application Information:
A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Beta Lipotropin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple epitopes because of the nature of the immunogen
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat