To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Tyrosine Kinase Receptor B
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Antigen symbol:TRKB
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µL
- Epitope:Rat mature TrkB protein (aa: 23-36), equivalent to aa 54-67 of full-length TrkB including signal peptide
- Cross adsorption:No
- Format:Lyophilized
- Antigen synonyms:Tropomyosin-related kinase receptor|Trkb
- Immunogen:A synthetic peptide (AFPRLEPNSIDPENC) as part of rat mature TrkB protein (aa: 23-36; equivalent to aa 54-67 of full-length TrkB) conjugated to KLH
- Cat. no.:10782-430
- Supplier No.:R-121-100
Specifications
About this item
FUNCTION: Receptor for brain-derived neurotrophic factor (BDNF), neurotrophin-3 and neurotrophin-4/5 but not nerve growth factor (NGF). Involved in the development and/or maintenance of the nervous system. This is a tyrosine-protein kinase receptor. Known substrates for the TRK receptors are SHC1, PI-3 kinase, and PLC-gamma-1. CATALYTIC ACTIVITY: ATP + a [protein]-L-tyrosine = ADP + a [protein]-L-tyrosine phosphate. SUBUNIT: Exists in a dynamic equilibrium between monomeric (low affinity) and dimeric (high affinity) structures (By similarity). Binds APS. Interacts with SQSTM1. SUBCELLULAR LOCATION: Membrane; single-pass type I membrane protein. ALTERNATIVE PRODUCTS: 3 named isoforms produced by alternative splicing. Additional isoforms seem to exist. TISSUE SPECIFICITY: The different forms are differentially expressed in various cell types. Isoform T2 is primarily expressed in neurons. PTM: Ligand-mediated auto-phosphorylation. SIMILARITY: Belongs to the Tyr protein kinase family. Insulin receptor subfamily. SIMILARITY: Contains 2 Ig-like C2-type (immunoglobulin-like) domains. SIMILARITY: Contains 2 LRR (leucine-rich) repeats. SIMILARITY: Contains 1 protein kinase domain.
Sequence was chosen to avoid cross reactivity with TrkA and TrkC.
Application Information:
IHC. A dilution of 1:1000 to 1:3000 is recommended for this application. The sequence of the immunogen and the conjugate are identical to the one used by Yan et al (see the Refs) and the antiserum appears to have similar characteristics. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: the Tyrosine Kinase Receptor B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: rat mature TrkB protein (aa: 23-36)
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat