To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Brain-Derived Neurotrophic Factor
- Clonality:Polyclonal
- Gene ID:P23560 BDNF_Human
- Host:Rabbit
- Isotype:IgG
- Neutralisation:Yes
- Reactivity:Human,Rat,Mouse
- Antigen symbol:BDNF
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µL
- Western blot:Yes
- Cross adsorption:No
- Format:Antiserum
- Form:Lyophilized
- Antigen synonyms:Brain-derived neurotrophic factor|proBDNF|Abrineurin
- Antibody fragment:IgG
- Storage temperature:After reconstitution keep aliquots at –20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Avoid repetitive freeze/thaw cycles. Glycerol (1:1) may be added for an additional stability.
- Shipping temperature:Ambient
- Immunogen:Recombinant human BDNF
- Purification:Whole serum
- Cat. no.:10782-374
- Supplier No.:R-088-100
Specifications
About this item
BDNF belongs to the neurotrophin family and regulates the survival and differentiation of neurons during development. The alterations in BDNF expression induced by various kinds of brain insult including stress, ischemia, seizure activity and hypoglycemia, may contribute to some pathologies such as depression, epilepsy, Alzheimer's, and Parkinson's disease. Microglia release BDNF that may contribute to neuroinflammation and neuropathic pain. FUNCTION: Promotes the survival of neuronal populations that are all located either in the central nervous system or directly connected to it. Major regulator of synaptic transmission and plasticity at adult synapses in many regions of the CNS. The versatility of BDNF is emphasized by its contribution to a range of adaptive neuronal responses including long-term potentiation (LTP), long-term depression (LTD), certain forms of short-term synaptic plasticity, as well as homeostatic regulation of intrinsic neuronal excitability. SUBUNIT: Monomers and homodimers. Binds to NTRK2/TRKB. SUBCELLULAR LOCATION: Secreted protein. Post Translation Modification (PTM): The propeptide is N-glycosylated and glycosulfated. PTM: Converted into mature BDNF by plasmin (PLG) (By similarity). DISEASE: Defects in BDNF are a cause of congenital central hypoventilation syndrome (CCHS); also known as congenital failure of autonomic control or Ondine curse. CCHS is a rare disorder characterized by abnormal control of respiration in the absence of neuromuscular or lung disease, or an identifiable brain stem lesion. A deficiency in autonomic control of respiration results in inadequate or negligible ventilatory and arousal responses to hypercapnia and hypoxemia. CCHS is frequently complicated with neurocristopathies such as Hirschsprung disease that occurs in about 16% of CCHS cases. SIMILARITY: Belongs to the NGF-beta family.
Less than 0.1% cross-reactivity against NGF, NT3 and NT4/5 by dot blot.
Application Information:
IHC, ELISA (1 site), Western Blot, inhibition of biological activity in vitro/in vivo. Recommended to be used at a dilution of 1:1000 for immunohistochemistry, ELISA and Western blot. 1:10 to 1:50 for inhibition of biological activity in vitro. Use neat for in vivo studies at 5-10 µl/g body weight. This antiserum stains cell bodies and some nerve terminals in the dorsal horn of the rat spinal cord, however, does not stain finest nerve terminals. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
IHC, ELISA (1 site), Western Blot, inhibition of biological activity in vitro/in vivo. Recommended to be used at a dilution of 1:1000 for immunohistochemistry, ELISA and Western blot. 1:10 to 1:50 for inhibition of biological activity in vitro. Use neat for in vivo studies at 5-10 µl/g body weight. This antiserum stains cell bodies and some nerve terminals in the dorsal horn of the rat spinal cord, however, does not stain finest nerve terminals.
Western Blotting: Antibody does detect BDNF forms in tissue lysates but there are multiple bands present, many of which are uncharacterized. The antibody detects 14 kDa (mature BDNF), 32 kDa (proBDNF) and a 18 kDa BDNF isoform (see blot examples). In cell lysates, only 18 kDa and 32 kDa BDNF are detected. The reason for these differences has not been characterized. Alternative antibodies for Western Blotting are: R-017-500 (IgG-purified form of R-088-100 for tissue homogenate analysis); R-1707-100 (cell lysates and tissue homogenates), R-083-100/R-066-500 (cell lysates, tissue homogenates and human serum); M-1744-50/100 (human serum and tissue homogenates).
Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: rh BDNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Mouse, Human