To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:pan Synuclein
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat
- Antigen symbol:pan Synuclein
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µL
- Western blot:Yes
- Epitope:amino acids 11-26 of Human alpha-, beta-, and gamma synuclein proteins
- Cross adsorption:No
- Format:Lyophilized
- Immunogen:A synthetic peptide (AKEGVVAAAEKTKQGV) as a consensus part of human alpha-, beta-, and gamma synuclein proteins conjugated to diphteria toxoid has been used as the immunogen.
- Cat. no.:10782-358
- Supplier No.:R-060-100
Specifications
About this item
NULL
Overlap specific immunohistochemical staining of alpha-, beta- and gamma synucleins
Application Information:
IHC, WB and immunoblot. A dilution of 1:1000 is recommended for these applications. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: pan synuclein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: amino acids 11-26 of Human alpha-, beta-, and gamma synuclein proteins
Host: Rabbit
Isotype: IgG
Reactivity: Rat