Order Entry
ContactUsLinkComponent
Anti-Myc Tag Mouse Monoclonal Antibody [clone: BMYCA7]
Anti-Myc Tag Mouse Monoclonal Antibody [clone: BMYCA7]
Catalog #: 10782-050
Supplier:  Biosensis
Antibody
Anti-Myc Tag Mouse Monoclonal Antibody [clone: BMYCA7]
Catalog #: 10782-050
Supplier:  Biosensis
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    Myc Epitope Tag
  • Clonality:
    Monoclonal
  • Clone:
    BMYCA7
  • Host:
    Mouse
  • Antigen symbol:
    Myc
  • Conjugation:
    Unconjugated
  • ImmunoPrecipitation:
    Yes
  • Size:
    100 µg
  • Western blot:
    Yes
  • Epitope:
    EQKLISEEDL
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Antigen synonyms:
    MRTL|bHLHe39|c-Myc
  • Immunogen:
    A synthetic peptide (EQKLISEEDL) coupled to KLH.
  • Cat. no.:
    10782-050
  • Supplier No.:
    M-1314-100

Specifications

About this item

The Myc tag contains the amino acids Glu-Gln-Lys-Leu-Ile-Ser-Glu-Glu-Asp-Leu (E-Q-K-L-I-S-E-E-D-L) corresponding to amino acids 410-419 of human Myc. This tag is widely used for monitoring expression of recombinant proteins in bacteria, insect and mammalian cells.

Antibody detects over-expressed proteins containing Myc epitope tag fused to either amino- or carboxy-termini of targeted proteins in transfected mammalian cells.
Application Information:
Western Blotting (WB), Immunoprecipitation (IP). Suggested starting dilutions as follows: WB at 1:1000, IP at 1:200. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Tag
Antigen: Myc Tag
Clonality: Monoclonal
Clone: BMYCA7
Conjugation: Unconjugated
Epitope: EQKLISEEDL
Host: Mouse
Isotype:
Reactivity:

Customers Who Bought This Also Bought