To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Low-affinity Nerve Growth Factor Receptor
- Clonality:Monoclonal
- Clone:ME20.4
- Host:Mouse
- Reactivity:Pig,Rabbit,Sheep,Human,Dog,Cat
- Antigen symbol:NGFR
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µg
- Epitope:The p75NTR antibody was derived from immunization of mice with human WM245 melanoma cells.
- Cross adsorption:No
- Format:Lyophilized
- Antigen synonyms:p75NTR|p75(NTR)|CD271|nerve growth factor receptor|Gp80-LTNFRSF16
- Immunogen:The p75NTR antibody was derived from immunization of mice with human WM245 melanoma cells.
- Cat. no.:10782-028
- Supplier No.:M-011-100
Specifications
About this item
Nerve growth factor receptor (NGFR) is also referred to as p75(NTR) due to its molecular mass and its ability to bind at low affinity not only NGF (see 162030), but also other neurotrophins, including brain-derived neurotrophic factor (BDNF; 113505), neurotrophin-3 (NTF3; 162660), and neurotrophin-4/5 (NTF5; 162662). At the time of its discovery, NGFR was considered a unique type of protein. Subsequently, however, a large superfamily of tumor necrosis factor receptors were found to share the overall structure of NGFR (4 extracellular ligand-binding, cysteine-rich repeats, or CRs, and signaling through association with, or disassociation from, cytoplasmic interactors). The identification of this superfamily helped elucidate some of the biologic functions of NGFR, including its ultimate involvement in the nuclear factor kappa-B (NFKB; see 164011) and apoptosis pathways. As a monomer, NGFR binds NGF with low affinity. Higher affinity binding is achieved by association with higher molecular mass, low-affinity neurotrophin receptors, namely the tropomyosin receptor kinases, TRKA (NTRK1; 191315), TRKB (NTRK2; 600456), and TRKC (NTRK3; 191316). TRKA, TRKB, and TRKC are specific for or 'preferred by' NGF, NTF5 and BDNF, and NTF3, respectively (Ip et al., 1993). NTF3 also binds to TRKA and TRKB, but with significantly lower affinity
This antibody recognises p75NTR (low affinity neurotrophin receptor)
Application Information:
Immunohistochemistry, immunofluorescence, flow cytometry. Suggested working dilutions: For Immunohistochemistry a concentration of 2 µg/ml is recommended. Antibody not appropriate for Western Blot. For FACS a concentration of 20 µg/ml is recommended. At least 1 in 5000 dilution is recommended for 1 site ELISAs. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: p75NTR
Clonality: Monoclonal
Clone: ME20.4
Conjugation: Unconjugated
Epitope: Human NT3 (19-128)
Host: Mouse
Isotype:
Reactivity: Human, Cat, Dog, Pig, Rabbit, Sheep