To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Adrenoceptor Beta 3
- Clonality:Polyclonal
- Host:Chicken
- Isotype:IgY
- Reactivity:Rat,Mouse
- Antigen symbol:ADRB3
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 µL
- Western blot:Yes
- Epitope:Mouse beta 3 Adrenergic Receptor (386-400 aa).
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:Adrb-3|FLJ99960|BETA3AR
- Immunogen:A synthetic peptide from mouse beta 3 Adrenergic Receptor (386-400 aa).
- Cat. no.:10781-956
- Supplier No.:C-1529-100
Specifications
About this item
Beta-adrenergic receptors are multi-pass membrane proteins that belong to the G-protein coupled receptor 1 family. Their function is to mediate the catecholamine-induced activation of adenylate cyclase through the action of G-proteins (ref: SWISSPROT).
This antibody detects ADRB3 in mouse and rat at approx 44 kDa.
Application Information:
WB and ELISA. Suggested dilution of 1:2,000-1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: ADRB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Mouse beta 3 Adrenergic Receptor (386-400 aa).
Host: Chicken
Isotype: IgY
Reactivity: Mouse, Rat