To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Fatty acid-binding Protein
- Clonality:Polyclonal
- Host:Chicken
- Isotype:IgY
- Reactivity:Human
- Antigen symbol:Fatty acid-binding Protein
- Conjugation:Unconjugated
- Size:100 µL
- Western blot:Yes
- Epitope:Maps to amino acids 97-111 of Human FABP4 protein
- Cross adsorption:Yes
- Format:Liquid
- Antigen synonyms:ALBP|Fatty acid-binding protein 4|FABP4
- Immunogen:Human Fatty acid-binding protein, adipocyte peptide (97-111 aa).
- Cat. no.:10781-952
- Supplier No.:C-1527-100
Specifications
About this item
Fatty acid-binding protein, adipocyte (ALBP) is a lipid transport protein which binds long chain fatty acids and other hydrophobic ligands and delivers them to their receptors in the nucleus. ALBP is found in the cytoplasm and nucleus of adipocytes.
This antibody detects human Fatty acid-binding protein, adipocyte at approx 16 kDa. The antibody may detect ALBP from other species because of protein homology
Application Information:
WB. Suggested dilution of 1:2,000-1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: Fatty acid-binding Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Maps to amino acids 97-111 of Human FABP4 protein
Host: Chicken
Isotype: IgY
Reactivity: Human