To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Polyclonal
- Host:Chicken
- Isotype:IgY
- Reactivity:Human,Rat,Mouse
- Antigen symbol:CNTF
- Conjugation:Unconjugated
- ELISA:Yes
- Size:500 µL
- Western blot:Yes
- Epitope:11-25 aa and 186-200 aa
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:Ciliary neurotrophic factor|CTNF
- Immunogen:Mixture of two peptides from human CNTF (11-25 and 186-200 aa). Both peptides are highly conserved among human and mouse.
- Cat. no.:10781-950
- Supplier No.:C-1526-500
Specifications
About this item
CNTF is a survival promoting factor for different types of neurons in vitro and in vivo. The essential structural features for the biological function of human CNTF were investigated by Thier, M. et al. They showed that deletion of 14 N-terminal and 18 C-terminal amino acids significantly increased bioactivity compared to wild-type CNTF. FUNCTION: CNTF is a survival factor for various neuronal cell types. Seems to prevent the degeneration of motor axons after axotomy. SUBUNIT: Homodimer. SUBCELLULAR LOCATION: Cytoplasm. TISSUE SPECIFICITY: Nervous system. PHARMACEUTICAL: CNTF is being tested under the name Axokine by Regeneron Pharmaceuticals for treatment of human motor neuron diseases, such as amyotrophic lateral sclerosis (ALS). As it induces substantial weight loss, preferentially of fat as opposed to lean body mass, it is being used for obesity treatment. SIMILARITY: Belongs to the CNTF family.
This antibody detects CNTF from human, rat, and mouse extracts in western blot and ELISA. Because of homology it is expected to react with other mammalian species as well. The antibody has not been tested in IHC
Application Information:
WB and ELISA at a suggested dilution of 1:500 to 1:2,000. A titration between 1:50 to 1:500 is suggested for IHC. Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: CNTF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: 11-25 aa and 186-200 aa
Host: Chicken
Isotype:
Reactivity: