To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Rhodopsin
- Clonality:Polyclonal
- Host:Chicken
- Isotype:IgY
- Reactivity:Tiger finch,Zebra finch,Rat,Mouse
- Antigen symbol:Rhodopsin
- Conjugation:Unconjugated
- Size:500 µL
- Western blot:Yes
- Epitope:Peptides cooresponding to amino acids 14-26 of Mouse Opsin 1 and amino acids 18-30 of Tiger finch opsin1.
- Cross adsorption:Yes
- Format:Liquid
- Antigen synonyms:opsin1|Red cone|Rho|Green cone
- Immunogen:Mixture of synthetic peptide (14-26 aa) of Mouse Green cones Opsin 1 and synthetic peptide (18-30 aa) of Tiger finch red cones opsin 1 was used as immunogen.
- Cat. no.:10781-940
- Supplier No.:C-1521-500
Specifications
About this item
Rod shaped photoreceptor cells that are required for image-forming vision at low light intensity and for photoreceptor cell viability after birth (ref: SWISSPROT).
The antibody reacts in wester blots in mouse brain extracts with a prominent band ~36kDa cooresponding to Rhodopsin. The antibody has not been tested in IHC.
Application Information:
WB. Suggested dilution at 1:500 to 1:2,000. To minimise background staining, a higher concentration of detergent (such as Tween-20) is suggested in the dilution and washing steps. Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: Rhodopsin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Peptides cooresponding to amino acids 14-26 of Mouse Opsin 1 and amino acids 18-30 of Tiger finch opsin1.
Host: Chicken
Isotype: IgY
Reactivity: Mouse, Rat, Tiger Finch, Zebra Finch