To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Neurotrophin 4
- Clonality:Polyclonal
- Host:Chicken
- Isotype:IgY
- Reactivity:Human,Mouse
- Antigen symbol:NTF4
- Conjugation:Unconjugated
- ELISA:Yes
- Size:500 µL
- Epitope:Human NT4/5 peptides, amino acids 104-118 and 198-210
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:Neutrophic factor 5|NTF5|Neurotrophin-4|NT-5|Neutrophic factor 4|Neurotrophin-5|NT-4|NTF4
- Immunogen:Mixture of two human NT4/NT5 peptides (104-118 and 198-210 aa).
- Cat. no.:10781-926
- Supplier No.:C-1514-500
Specifications
About this item
NT4 is a member of the neurotrophin family, in knockout animals NT4 deficient mice appeared little changed from wildtype animals, however NT4 is a powerful neurotrophic factor for periferial neuronal ganglia and is capable if sustaining ganglia neurons in vitro. NT4 is closely related to both NGF and BDNF. FUNCTION: Target-derived survival factor for peripheral sensory sympathetic neurons. SUBCELLULAR LOCATION: Secreted protein. TISSUE SPECIFICITY: Highest levels in prostate, lower levels in thymus, placenta, and skeletal muscle. Expressed in embryonic and adult tissues. SIMILARITY: Belongs to the NGF-beta family.
The antibody reacts with human and mouse NT-4//5 proteins and likely with other species based upon homology. The antibody has not been tested in IHC or western blot
Application Information:
ELISA at suggested dilution of 1:500-1:2,000. Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: Neurotrophin-4/5; NT4/NT5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human NT4/5 peptides, amino acids 104-118 and 198-210
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse