To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Neurotrophin 3
- Clonality:Polyclonal
- Host:Chicken
- Isotype:IgY
- Reactivity:Human,Mouse
- Antigen symbol:NTF3
- Conjugation:Unconjugated
- ELISA:Yes
- Size:500 µL
- Epitope:Human NT3 peptides, amino acids 90-104 and 199-214.
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:NGF-2|HDNF|Ntf3|Neurotrophin-3|Nerve growth factor 2|Ntf-3|Neurotrophic factor
- Immunogen:Mixture of two human NT3 peptides (90-104 and 199-214 aa).
- Cat. no.:10781-924
- Supplier No.:C-1513-500
Specifications
About this item
NT3 is a member of the neurotrophin family, that controls survival and differentiation of visceral and proprioceptive sensory neurons. NT3 is closely related to both NGF and BDNF. It may be involved in the maintenance of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NT3-deficient mice generated by gene targeting display sevvere movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse. SUBCELLULAR LOCATION: Secreted protein. TISSUE SPECIFICITY: Brain and peripheral tissues. SIMILARITY: Belongs to the NGF-beta family
The antibody reacts with human and mouse NT-3 proteins and likely with other species based upon homology. The antibody has not been tested in IHC or western blot
Application Information:
ELISA at suggested dilution of 1:500-1:2,000. Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: Neurotrophin-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human NT3 peptides, amino acids 90-104 and 199-214.
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse