Determination of the Properties of Buffer Solutions
Catalog #: 470163-112
Supplier: Ward's Science
Specifications
- Description:Determination of the Properties of Buffer Solutions
- No. of Lab Groups:15
- Cat. no.:470163-112
- Includes:Teacher's Guide and Student Copymaster
- Supplier No.:IS8024
Specifications
About this item
What Does A Buffer Do?
- Classic AP Chemistry Experiment
- Instructions Included
In this experiment, you will prepare three buffer solutions having different pH values and show that the pH of these solutions does not change significantly when small amounts of acids and bases are added. You will also show that when the same amounts of acids and bases are added to water and to a non-buffer solution (e.g. NaCl solution), the pH changes are large.
Includes:
Sodium Acetate, trihydrate
Ammonium Chloride
Sodium Bicarbonate
Sodium Chloride
Acetic Acid EZ-Prep
Ammonium Hydroxide EZ-Prep
Sodium Hydroxide Solution
Hydrochloric Acid Solution
Customers Who Bought This Also Bought