Vacuum Lifter
Catalog #: 470163-920
Supplier: American Scientific
Specifications
- Description:Vacuum Lifter
- Cat. no.:470163-920
- Dimensions:10.5 x 10.5"
- Supplier No.:AR-510
Specifications
About this item
Demonstrates Atmospheric Pressure.
- Explore additional concepts such as surface area, force, vacuums, and pressure
This durable, thick rubber mat has a simple design. Pick up an ordinary flat lab stool or make a challenge to lift off a table, which is nearly impossible! Show how suction cups stay stuck because of the pushing force of air, not from a pulling "suction" force. Comes with instructions.
Customers Who Bought This Also Bought